Infrared optical windows
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
38384.KT 38391.KT 38392.KT 38393.KT 38395.KT 38397.KT 38399.KT 38413.KT 38414.KT 38415.KT 38416.KT 38417.KT 38420.KT 38423.KT 38425.KT 38426.KT 38427.KT 38428.KT 38429.KT 38430.KT 38433.KT 38435.KT 38436.KT 38438.KT 38443.KT 38445.KT 38446.KT 38447.KT 38449.KT 38451.KT 38452.KT 38453.KT 38454.KT 38455.KT 38457.KT 38458.KT 38461.KT 38467.KT 38469.KT 38470.KT 38473.KT 38476.KT 38478.KT 38481.KT 41740.KT 41744.KT 41883.KT 42145.KT 42146.KT 45944.KT
38384.KTEA 43.7 EUR
38384.KT 38391.KT 38392.KT 38393.KT 38395.KT 38397.KT 38399.KT 38413.KT 38414.KT 38415.KT 38416.KT 38417.KT 38420.KT 38423.KT 38425.KT 38426.KT 38427.KT 38428.KT 38429.KT 38430.KT 38433.KT 38435.KT 38436.KT 38438.KT 38443.KT 38445.KT 38446.KT 38447.KT 38449.KT 38451.KT 38452.KT 38453.KT 38454.KT 38455.KT 38457.KT 38458.KT 38461.KT 38467.KT 38469.KT 38470.KT 38473.KT 38476.KT 38478.KT 38481.KT 41740.KT 41744.KT 41883.KT 42145.KT 42146.KT 45944.KT
Infrared optical windows
Optische ramen
These optical crystal windows are ideal for infrared spectroscopy.
  • Disc or rectangular
  • Polished or unpolished
A polishing kit is available for those who wish to polish their windows.