Trousers, PW3, T602 Urban Work Holster
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T602BKR46 T602BKR36 T602BKR32 T602BKR38 T602BKR41 T602BKR40 T602BKR44 T602BKR34 T602BKR33 T602BKR42 T602BKR48 T602BKR28 T602BKR30 T602ZBR28 T602ZBR30 T602ZBR32 T602ZBR33 T602ZBR40 T602ZBR34 T602ZBR36 T602ZBR38 T602ZBR44 T602ZBR42 T602ZBR41 T602ZBR46 T602ZBR48
PRTWT602BKR46EA 54.8 EUR
PRTWT602BKR46 PRTWT602BKR36 PRTWT602BKR32 PRTWT602BKR38 PRTWT602BKR41 PRTWT602BKR40 PRTWT602BKR44 PRTWT602BKR34 PRTWT602BKR33 PRTWT602BKR42 PRTWT602BKR48 PRTWT602BKR28 PRTWT602BKR30 PRTWT602ZBR28 PRTWT602ZBR30 PRTWT602ZBR32 PRTWT602ZBR33 PRTWT602ZBR40 PRTWT602ZBR34 PRTWT602ZBR36 PRTWT602ZBR38 PRTWT602ZBR44 PRTWT602ZBR42 PRTWT602ZBR41 PRTWT602ZBR46 PRTWT602ZBR48
Trousers, PW3, T602 Urban Work Holster
Werkkleding
Rugged trouser made from durable Kingsmill polyester/cotton 300 g fabric with a modern fit and adjustable leg length. Pockets have oxford reinforcement for extra durability.
  • Gripper elasticated inner waistline helping keep upper body garments in place
  • Side elastic waist for ultimate wearer comfort
  • Adjustable hem to accommodate all leg lengths
  • Reinforced panels in high wear areas for maximum durability
  • Contrast panels for protection against dirt
  • Triple-stitched seams for extra durability
  • Contrast colour stitching for added style
  • 11 pockets for ample storage including holster pockets, top loading kneepad pockets, side leg cargo pocket
Shell Fabric: Kingsmill 300 g
Contrast Fabric: 100% Polyester 600D Fabric 210 g
Certificaten: EN 14404:2004+A1:2010 Type 2 (Level 0 (When used in combination with S156)))