Aquet detergent for glassware and plastics, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F170940020 F170940050
BELAF170940020EA 163 EUR
BELAF170940020 BELAF170940050
Aquet detergent for glassware and plastics, SP Bel-Art
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Safe for glassware, plastics, and surfaces, this phosphate-free, nonionic surfactant will not etch or mar precision glassware. It rinses quickly and dries without wiping.
  • Convenient, pre-portioned pouches make one gallon of 1% solution
  • Not for use in automatic dishwashers