Petri dish platinum electrodes
Leverancier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45-2059 45-2060
BTXI45-2059EA 3260 EUR
BTXI45-2059 BTXI45-2060
Petri dish platinum electrodes
Elektroden Elektroden voor elektroporatie
These reusable oocyte Petri electrodes are ideal for electroporation of oocytes, embryos, or other cell types in small volumes.
  • Electrodes are permanently attached to a glass Petri dish and come with a glass lid. Designed to easily fit on an inverted microscope stage to allow easy observation of cells during electroporation.