ESD safety shoes, lace-up, L10, Shadow
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
721081/37 721081/38 721081/39 721081/40 721081/41 721081/42 721081/43 721081/44 721081/45 721081/46 721081/47
114-8738EA 90.9 EUR
114-8738 114-8739 114-8740 114-8741 114-8742 114-8743 114-8744 114-8745 114-8746 114-8747 114-8748
ESD safety shoes, lace-up, L10, Shadow
Schoenen en laarzen
Casual, vintage style ESD safety shoes for ladies made from cowhide leather, a padded tongue plus a low profile, dual-density TPU/PU sole (L10).
  • Leather lining
  • Anatomically-shaped insole supports natural heel-to-toe foot movement
  • Full length inlay sole (ESD Pro), beige
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.