ESD safety shoes, sandals, Velcro® fastening, Sid
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
72131/38 72131/39 72131/40 72131/41 72131/42 72131/43 72131/44 72131/45 72131/46 72131/47
ELTN72131/38EA 81.9 EUR
ELTN72131/38 ELTN72131/39 ELTN72131/40 ELTN72131/41 ELTN72131/42 ELTN72131/43 ELTN72131/44 ELTN72131/45 ELTN72131/46 ELTN72131/47
ESD safety shoes, sandals, Velcro® fastening, Sid
Schoenen en laarzen
100% metal- and leather-free, sporty and lightweight ESD safety shoe made from hydrophobised microfibre and hydrophobised Cordura® (tear and abrasion resistant, dirt and water repellent), a padded tongue and a slip resistant dual-density TPU/PU sole Trainers.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length fleece inlay sole ESD PRO black
  • TPU scuff cap protects the upper
  • Metal-free, puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC