ESD safety shoes, lace-up, L10, Vintage Pirate
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
722071/37 722071/38 722071/39 722071/40 722071/41 722071/42 722071/43 722071/44 722071/45 722071/46 722071/47
114-8749EA 90.9 EUR
114-8749 114-8750 114-8751 114-8752 114-8753 114-8754 114-8755 114-8756 114-8757 114-8758 114-8759
ESD safety shoes, lace-up, L10, Vintage Pirate
Schoenen en laarzen
Vintage and casual style, environmental friendly manufactured ESD safety shoe made from waxy cowhide leather, a padded tongue and a low profile dual-density VIBRAM® rubber/PU tread sole L10 PIRATE.
  • Fabric lining Bioactive
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO grey
  • Anatomically shaped insole and ankle support for improved stability
  • Abrasion resistant scuff cap
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Grey/red
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC