ESD safety shoes, slip-on, White Slipper
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
72498/45 72498/46 72498/48 72498/43 72498/44 72498/47 72498/41 72498/42 72498/39 72498/40
ELTN72498/45EA 72.9 EUR
ELTN72498/45 ELTN72498/46 ELTN72498/48 ELTN72498/43 ELTN72498/44 ELTN72498/47 ELTN72498/41 ELTN72498/42 ELTN72498/39 ELTN72498/40
ESD safety shoes, slip-on, White Slipper
Schoenen en laarzen
Leather-free ESD safety shoe made from hydrophobised microfibre plus a mono-density sole (Safety-Grip).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), grey
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: White
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC.