ESD safety shoes, lace-up, Journey S1P
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
729621/40 729621/41 729621/42 729621/43 729621/44 729621/45 729621/46 729621/47 729621/48
ELTN729621/40EA 85.9 EUR
ELTN729621/40 ELTN729621/41 ELTN729621/42 ELTN729621/43 ELTN729621/44 ELTN729621/45 ELTN729621/46 ELTN729621/47 ELTN729621/48
ESD safety shoes, lace-up, Journey S1P
Schoenen en laarzen
Sporty ESD safety shoes made from suede, mesh and microfibre upper, a skin friendly textile lining, a padded tongue plus a dual-density rubber/PU sole (E-Track).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (Sportive ESD)
  • Soft fleece ESD insole
  • Anatomically-shaped insole and ankle support for improved stability
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/blue or blue/red
Certificaten: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC.