ESD safety shoes, lace-up, Nova, Maja S3
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74781/35 74781/36 74781/37 74781/38 74781/39 74781/40 74781/41 74781/42
ELTN74781/35EA 80.9 EUR
ELTN74781/35 ELTN74781/36 ELTN74781/37 ELTN74781/38 ELTN74781/39 ELTN74781/40 ELTN74781/41 ELTN74781/42
ESD safety shoes, lace-up, Nova, Maja S3
Schoenen en laarzen
Leather-free, lightweight ESD safety shoe made from microfibre/textile upper, a padded tongue and collar plus a slip resistant, dual-density PU/PU sole Nova.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole Ladies ESD
  • Soft fleece ESD insole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.