ESD safety ankle boots, lace-up, Nova, Maja
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74791/37 74791/38 74791/35 74791/36 74791/39 74791/40 74791/41 74791/42
ELTN74791/37EA 82.9 EUR
ELTN74791/37 ELTN74791/38 ELTN74791/35 ELTN74791/36 ELTN74791/39 ELTN74791/40 ELTN74791/41 ELTN74791/42
ESD safety ankle boots, lace-up, Nova, Maja
Schoenen en laarzen
Leather-free, lightweight ESD safety ankle boot made from microfibre/textile upper, a padded tongue and collar and a slip resistant dual-density PU/PU sole NOVA.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole LADIES ESD
  • Soft fleece ESD insole
  • Puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC