ESD safety ankle boots, lace-up, L10, Sensation
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
76106/37 76106/38 76106/39 76106/40 76106/41 76106/42 76106/43 76106/44 76106/45 76106/46 76106/47 76106/48
ELTN76106/37EA 84.9 EUR
ELTN76106/37 ELTN76106/38 ELTN76106/39 ELTN76106/40 ELTN76106/41 ELTN76106/42 ELTN76106/43 ELTN76106/44 ELTN76106/45 ELTN76106/46 ELTN76106/47 ELTN76106/48
ESD safety ankle boots, lace-up, L10, Sensation
Schoenen en laarzen
Casual, vintage style, environmental friendly manufactured ESD safety shoe made from hydrophobised textile 'denim look' material and hydrophobised nubuck leather, a padded tongue plus a low profile, dual-density TPU/PU sole (L10).
  • Bioactive fabric lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), grey
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/grey
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC.