ESD safety ankle boots, lace-up, Biomex Dynamics, Tommaso Green
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
765501/40 765501/41 765501/42 765501/43 765501/44 765501/45 765501/46 765501/47 765501/48
ELTN765501/40EA 117.9 EUR
ELTN765501/40 ELTN765501/41 ELTN765501/42 ELTN765501/43 ELTN765501/44 ELTN765501/45 ELTN765501/46 ELTN765501/47 ELTN765501/48
ESD safety ankle boots, lace-up, Biomex Dynamics, Tommaso Green
Schoenen en laarzen
Metal- and leather-free, lightweight ESD safety shoe made from perforated microfibre and reflective materials, skin friendly textile lining, a full padded bellows tongue and collar plus an extremely flexible, ergonomic S-shaped, dual-density TPU/PU sole (Biomex Dynamics).
  • Climate-regulating, breathable textile lining
  • Anatomically-shaped insole supports the natural heel-to-toe foot movement
  • Replaceable insole; orthopaedic insoles possible
  • Full length ESD inlay sole
  • Biomex-Protection®: Cuff stabilises ankles
  • Abrasion-proof scuff cap
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/green
Certificaten: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC.