ESD safety ankle boots, lace-up, Sander
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
76872/35 76872/36 76872/37 76872/38 76872/41 76872/42 76872/43 76872/44 76872/45 76872/46 76872/47 76872/48 76872/39 76872/40
ELTN76872/35EA 84.9 EUR
ELTN76872/35 ELTN76872/36 ELTN76872/37 ELTN76872/38 ELTN76872/41 ELTN76872/42 ELTN76872/43 ELTN76872/44 ELTN76872/45 ELTN76872/46 ELTN76872/47 ELTN76872/48 ELTN76872/39 ELTN76872/40
ESD safety ankle boots, lace-up, Sander
Schoenen en laarzen
100% metal- and leather-free, lightweight ESD safety ankle boot made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black/red
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC