ESD safety ankle boots, lace-up, MADDOX
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
769561/46 769561/40 769561/43 769561/44 769561/45 769561/47 769561/48 769561/41 769561/42
ELTN769561/46EA 93.9 EUR
ELTN769561/46 ELTN769561/40 ELTN769561/43 ELTN769561/44 ELTN769561/45 ELTN769561/47 ELTN769561/48 ELTN769561/41 ELTN769561/42
ESD safety ankle boots, lace-up, MADDOX
Schoenen en laarzen
Leather-free ESD safety ankle boots made from hydrophobised microfibre and hydrophobised textile material, a full padded bellows tongue plus a dual-density TPU/PU sole (Maxximo).
  • Climate-regulating, breathable textile lining
  • Full length inlay sole (Sportive ESD)
  • Cushioned midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving energy back to the wearer
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certificaten: EN 61340-5-1 (ESD); EN ISO 20345; S3, SRC.