ESD safety ankle boots, lace-up, Sander XXT
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
769731/36 769731/37 769731/38 769731/39 769731/40 769731/41 769731/42 769731/43 769731/44 769731/45 769731/46 769731/47 769731/48 769731/35 769731/49
114-8435EA 97.9 EUR
114-8435 114-8436 114-8437 114-8438 114-8439 114-8440 114-8441 114-8442 114-8443 114-8444 114-8445 114-8446 114-8447 ELTN769731/35 ELTN769731/49
ESD safety ankle boots, lace-up, Sander XXT
Schoenen en laarzen
100% metal- and leather-free, lightweight ESD safety ankle boot made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), a padded tongue plus a slip resistant, dual-density TPU/PU sole (Trainers).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • TPU scuff cap protects the leather upper
  • Metal-free puncture resistant midsole
  • Composite toe protection cap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/red
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC