ESD safety rigger boots, pull-on, Safety-Grip, Will
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86771/36 86771/37 86771/38 86771/39 86771/40 86771/41 86771/42 86771/43 86771/44 86771/45 86771/46 86771/47 86771/48 86771/49 86771/50
ELTN86771/36EA 78.9 EUR
ELTN86771/36 ELTN86771/37 ELTN86771/38 ELTN86771/39 ELTN86771/40 ELTN86771/41 ELTN86771/42 ELTN86771/43 ELTN86771/44 ELTN86771/45 ELTN86771/46 ELTN86771/47 ELTN86771/48 ELTN86771/49 ELTN86771/50
ESD safety rigger boots, pull-on, Safety-Grip, Will
Schoenen en laarzen
These ESD winter safety boots are made from cowhide leather, reflective materials and a PU/PU walking sole Safety Grip.
  • Warm lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length warm fleece ESD inlay sole
  • Anatomically shaped insole supports the natural heel-to-toe movement of the foot
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, CI