FMOC-O-tert-butyl-L-tyrosine 98% (by HPLC)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: N-α-Fmoc-o-t-butyl-L-Tyrosine, N-Fmoc-O-tert-butyl-L-tyrosine, Nalpha-Fmoc-O-tert-butyl-L-tyrosine, O-tert-Butyl-Nalpha-Fmoc-L-tyrosine, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine, Fmoc-Tyr(tBu)-OH
H59730.06 H59730.14
H59730.06EA 60.5 EUR
H59730.06 H59730.14
FMOC-O-tert-butyl-L-tyrosine 98% (by HPLC)
FMOC-O-tert-butyl-L-tyrosine
Formule: C₂₈H₂₉NO₅
MW: 459.54 g/mol
Smeltpunt: 153…156 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00037129
CAS nummer: 71989-38-3
EINECS: 276-262-7
Order Now
Specification Test Results
Assay (HPLC) >97.5%