Spill containment pools, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PAK925 PAK926 PAK927 PAK928
NEWPPAK925EA 137 EUR
NEWPPAK925 NEWPPAK926 NEWPPAK927 NEWPPAK928
Spill containment pools, PIG®
Absorptiematerialen en bescherming tegen morsen
These containment pools are ideal for quick deployment to catch the contents of blown hydraulic lines, punctured tanks, leaking containers and decontamination operations. To store our pop-up containment pool in the vehicle to be prepared for spill emergencies on the road.
  • Made of heavy-duty, 18 oz. vinyl-coated polyester provides high resistance to tears and punctures
  • Rim has foam ring that rises with level of liquid contained
  • Deploys instantly for use in an emergency; decontaminate for reuse
  • Pliable material folds easily for storage on a truck or