N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4001028.0005 4001028.0025
BACE4001028.0005EA 0 EUR
BACE4001028.0005 BACE4001028.0025
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride
Substrate for trypsin, papain, acrosin, plasmin, cathepsin B, and cathepsin H.
Formule: C₁₉H₂₃ClN₆O₄
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00012846
CAS nummer: 911-77-3
EINECS: 213-011-2
Order Now
Specificaties
Environmentally Preferable