Precision thermometers with straight stem
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L36223-TOL L36756-FL L36000-TOL L36020-CB L36044-CB L36074-CB L36122-CB L36146-FL L36206-TOL L36206-TOL/VC L36252-FL L36408-PENT L36712-FL L36716-FL L36716-FL/VC L36724-FL L36732-FL L36736-FL
AMARL36223-TOLEA 81.5 EUR
AMARL36223-TOL AMARL36756-FL AMARL36000-TOL AMARL36020-CB AMARL36044-CB AMARL36074-CB AMARL36122-CB AMARL36146-FL AMARL36206-TOL AMARL36206-TOL/VC AMARL36252-FL AMARL36408-PENT AMARL36712-FL AMARL36716-FL AMARL36716-FL/VC AMARL36724-FL AMARL36732-FL AMARL36736-FL
Precision thermometers with straight stem
Thermometers Analoge thermometers
These immersion thermometers are suitable for government verification and calibration.
  • Enclosed scale with milk glass incorporated in the glass tube
  • Prismatic capillary
  • Richter-top finish without button
Toluene, pentene, blue or red spirit.
Verpakking: Supplied in transparent square plastic cases.