TLC adsorbents, silica gel
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
816320.1 816310.1 816310.5 816320.5 816330.1 816330.5 816340.1 816340.5 816380.1 816380.5 816400.1 816400.5 816410.1 816410.5
MANA816320.1EA 95 EUR
MANA816320.1 MANA816310.1 MANA816310.5 MANA816320.5 MANA816330.1 MANA816330.5 MANA816340.1 MANA816340.5 MANA816380.1 MANA816380.5 MANA816400.1 MANA816400.5 MANA816410.1 MANA816410.5
TLC adsorbents, silica gel
Sorbenten voor chromatografie Sorbenten voor TLC
Silica adsorbents for thin layer chromatography.
  • Available in different quality grades
  • Adsorbents with or without binder
  • Available with or without fluorescence indicator
Pore volume: 0,75 ml/g
Specific surface (BET): 500 m²/g