TLC indicators
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
816710.01 816720.01
MANA816710.01EA 76.7 EUR
MANA816710.01 MANA816720.01
TLC indicators
Sorbenten voor chromatografie Sorbenten voor TLC
Fluorescent indicators for thin layer chromatography.
  • UV indicators with efficient radiation for short-wave as well as long-wave UV ranges
  • Can be used for preparation of TLC plates
UV254: Manganese activated zinc silicate with absorption maximum at 254 nm; green fluorescence.
UV366: Inorganic fluorescent pigment with absorption maximum at 366 nm; blue fluorescence.