TLC visualisation reagents
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
814919 814921 814923
MANA814919EA 43.3 EUR
MANA814919 MANA814921 MANA814923
TLC visualisation reagents
Sorbenten voor chromatografie Sorbenten voor TLC
Selection of spray reagents for detection reactions in TLC.
  • Postchromatographic detection
  • Suited for spraying or dipping TLC plates
Opgelet: These products contain harmful substances which must be specially labeled as hazardous.