TLC education material kit, micro-set M
Leverancier: Macherey-Nagel
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
552-0282EA 373 EUR
552-0282
TLC education material kit, micro-set M
Educatief materiaal Educatief materiaal over chemie Leeractiviteiten voor chromatografie
Materials kit for scientific education.
  • Materials for the advanced kits F1, F2 and F3
  • Also usable as basic equipment for individual TLC studies
  • Developing chambers, glass capillaries, measuring cylinder, TLC sheets etc. are included
For beginners TLC micro-set A is recommendable. It contains all chemicals and accessories for fascinating experiments.