TLC education advanced kit, micro-set F1
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
814200 814201 814202 814203 814204 814205 814206
552-0287EA 284 EUR
552-0287 MANA814201 MANA814202 MANA814203 MANA814204 MANA814205 MANA814206
TLC education advanced kit, micro-set F1
Educatief materiaal Educatief materiaal over chemie Leeractiviteiten voor chromatografie
Advanced kit for science education.
  • All chemicals included for the separation of:
  • Amino acids test mixture and amino acids in urine sample
  • Heavy metal cations
Together with the materials from TLC micro-Set M an amino acid mixture (alanine, arginine, tryptophan, valine) as well as an heavy metal cation mixture (copper(II), manganese(II)) can be separated and visualised by spray reagents (ninhydrin and rubeanic acid).
Opgelet: * These products contain harmful substances which must be specially labeled as hazardous.