Rhodium(III) chloride solution 10 - 30% w/w (cont, Rh) 99.5% (w/w)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Rhodium(III) chloride, Rhodium trichloride
46523.NJ 46523.NA
46523.NJEA 397 EUR
46523.NJ 46523.NA
Rhodium(III) chloride solution 10 - 30% w/w (cont, Rh) 99.5% (w/w)
Rhodium(III) chloride
Formule: Cl₃Rh
MW: 209.26 g/mol
Dichtheid: 5.38
MDL Nummer: MFCD00011204
CAS nummer: 10049-07-7
UN: 3264
ADR: 8 III
Merck Index: 14,8188