Cut resistant gloves, uvex phynomic C5
Leverancier: Uvex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
uvex phynomic
112-5372EA 140 EUR
112-5372 112-5373 112-5374 112-5375 112-5376 112-5377 112-5378
Cut resistant gloves, uvex phynomic C5
Handschoenen
These Dyneema®Diamond Technology/polyamide/elastane gloves feature a knitted cuff and an aqua-polymer foam coating on palm and fingertips that provides very good mechanical abrasion resistance. These safety gloves are like a second skin and are especially light and flexible – without making your hands immediately sweaty.
  • Good grip in dry and slightly damp areas
  • Excellent cut protection (level C) and high tear resistance thanks to optimum material composition
  • Very high level of breathability
  • Perfect fit with 3D Ergo technology
  • Excellent skin tolerance – confirmed by the proDERM Institute for Applied Dermatological Research
Suitable for use in precision assembly work, precision work, inspection, sorting and food processing.
Certificaten: EN 388:2016 (4X42C)
Food contact approved
OEKO-Tex® Standard 100