Alum adjuvant, AlumVax hydroxide
Leverancier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AH0050 AH0250
OZBIAH0050EA 119.9 EUR
OZBIAH0050 OZBIAH0250
Alum adjuvant, AlumVax hydroxide
Eiwitexpressie Reagentia voor eiwitexpressie
AlumVax hydroxide is a wet gel (colloidal) of aluminium hydroxide 2%. It is the most common adjuvant used in approved prophylactic vaccines because of its excellent safety profile and ability to enhance the protective humoral immune response.
  • Ready to use suspension
  • Store at room temperature
It has been observed that aluminium compounds act by a depot effect and also by direct activation of the immune cells. Adsorption or entrapment of antigens in aggregates through hydrophobic and electrostatic interactions favours a high local antigen concentration and improved uptake by antigen presentating cells (APC).
AlumVax hydroxide is a crystalline aluminium oxyhydroxide that is positively charged at physiological pH (pI=11), suitable for adsorption of negatively charged, acidic proteins (such as albumin). It stimulates Th2 response through the release of Th2-associated cytokines (IL4, IL-5, IL-13) and Th2-associated antibodies (IgG1 and IgE). It increases Ag-specific CD4+ T Cell proliferation and promotes NALP3 inflammasome activation and caspase 1-mediated release of IL-1 and IL-18.
Leveringsinformatie: Shipped at room temperature.
Opgelet: Intended for research use only.