Protein transfection reagent, Pro-DeliverIN CRISPR
Leverancier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIC60100 PIC60500 CAS9PIC
OZBIPIC60100EA 204 EUR
OZBIPIC60100 OZBIPIC60500 OZBICAS9PIC
Protein transfection reagent, Pro-DeliverIN CRISPR
Transfectiereagentia
Pro-DeliverIN CRISPR is a transfection reagent optimised for recombinant Cas9 protein delivery or Cas9/gRNA RNP complexes. Suitable for gene editing applications, this reagent provides high transfection efficiency with minimal toxicity.
  • High efficiency delivery of Cas9 protein or Cas9 gRNA complexes
  • Low cell toxicity - enable to start experiment with less cells
The Cas 9 recombinant protein is delivered more rapidly than nucleic acid and is fully active once inside the cells without latency period (in contrast to transcription and translation machineries required for the nucleic acids). As for mRNA, it also offers the advantage of avoiding genomic integration and is cell cycle independent.
Efficient delivery represents a critical step for genome editing experiments.
Leveringsinformatie: Supplied with 100 µl of R-Phycoerythrin positive control.