Guard column holder for preparative HPLC columns, VarioPrep
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
718251 718256 718253 718255
MANA718251EA 300 EUR
MANA718251 MANA718256 MANA718253 MANA718255
Guard column holder for preparative HPLC columns, VarioPrep
Houder voor bewakingskolom
VarioPrep (VP) guard column holders are needed for the installation of VP guard columns at VP preparative HPLC columns.
  • Ideal protection for your preparative main column
  • VP column system with minimised void volume
VP guard column are packed with NUCLEODUR® or NUCLEOSIL® adsorbents for HPLC.
Order Now
Specificaties
Environmentally Preferable