Squalene emulsion, SqualVax
Leverancier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SQ0010 SQ0050 SQ0100
OZBISQ0010EA 176.79 EUR
OZBISQ0010 OZBISQ0050 OZBISQ0100
Squalene emulsion, SqualVax
Eiwitexpressie Reagentia voor eiwitexpressie
SqualVax is an oil-in-water emulsion made of squalene droplets in a continuous aqueous phase. It is an important advantage over alternative oils that have been used in emulsion adjuvants, like Freund’s adjuvant that contains mineral oil (paraffin oil) and has long term persistence in organisms.
Squalene emulsion induces local stimulation and recruitment of DCs and granulocytes, differentiation of monocytes into DCs and increased uptake of antigen by DCs. The emulsion acts more specifically on macrophages present at the site of injection. This formulation enhances differentiation of monocytes towards a mature phenotype, thereby promoting migration of antigen-loaded cells to the draining lymph node.
Leveringsinformatie: Shipped at room temperature.
Verpakking: Store at +4 °C.
Order Now
Specificaties
Environmentally Preferable