ESD safety shoes, lace-up, Wellmaxx, Miles, Rob
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
729451/40 729451/41 729451/42 729451/43 729451/44 729451/45 729451/46 729451/47 729451/48
ELTN729451/40EA 107.9 EUR
ELTN729451/40 ELTN729451/41 ELTN729451/42 ELTN729451/43 ELTN729451/44 ELTN729451/45 ELTN729451/46 ELTN729451/47 ELTN729451/48
ESD safety shoes, lace-up, Wellmaxx, Miles, Rob
Schoenen en laarzen
Lightweight and comfortable ESD safety shoes made from cowhide leather/hydrophobized nubuck leather, full padded bellows tongue plus collar and a dual-density TPU/PU sole WELLMAXX.
  • Climate regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full-length inlay sole ESD PRO black
  • Soft fleece ESD insole
  • Scuff cap for additional protection
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • Puncture resistant steel midsole
  • Protective composite toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey
Certificaten: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC