Disposable culture tubes, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
261322108 261322305
391-0190EA 112 EUR
391-0190 391-0195
Disposable culture tubes, DURAN®
Buisjes Cultuurbuisjes
DURAN®, borosilicaatglas 3.3, helder.
  • High thermal resistance, autoclavable
  • Very good chemical and physical properties
Reusable culture tubes suitable for Kapsenberg caps.