Transduction enhancer, in vivo ViroMag™
Leverancier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IV-VM30250 IV-VM30500 IV-TK30230 IV-KC30230
OZBIIV-VM30250EA 263.55 EUR
OZBIIV-VM30250 OZBIIV-VM30500 OZBIIV-TK30230 OZBIIV-KC30230
Transduction enhancer, in vivo ViroMag™
Transfectiereagentia
In vivo ViroMag™ transfection reagent has been designed for in vivo targeted transduction with any viral vector. Virus/nanoparticle complexes can be easily administrated through various injection routes such as systemic administration (intravenous, intra-artery) or local administration (intratumoral, intracerebroventricular).
  • High and increased transduction/infection efficiency
  • Magnetically targeted transduction to specific area
  • Reduction of virus dose and systemic dissemination
  • Work under non permissive conditions (hypothermia, physiological flow conditions)
  • Penetration of the vector into tissues
  • Minimised toxicity
  • Suitable for all types of viruses
Store at +4 °C.
Bestelinformatie: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Leveringsinformatie: Shipped at room temperature.