Sample loop kit
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18-0404-01EA 470 EUR
18-0404-01
Sample loop kit
Benodigdheden voor chromatografie Injectieloops voor chromatografiesystemen
Each sample loop consists of tubing 1,8 mm OD and two tubing connectors (M6).
Leveringsinformatie: The kit contains: 1×25 µl sample loop 0,5 mm ID; 1×50 µl sample loop 0,5 mm ID; 1×100 µl sample loop 0,5 mm ID; 1×200 µl sample loop 0,8 mm ID; 1×500 µl sample loop 0,8 mm ID.