VWR®, Contact slides
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
525472VEA 32.3 EUR
525472V 525392V 525262V 525482V 525272V 525322V
VWR®, Contact slides
Media voor microbiologie Geprepareerde media voor microbiologie
Surface and liquid flexible slides for simple and effective microbiological monitoring and hygiene testing of surfaces and fluids.
  • Two tests with one slide: Each slide is coated on both sides either with the same agar or with a different agar on each side
  • Long shelf life: 9 months from production
  • Clear instructions allow a simple way to evaluate the microbiological contamination of fluid surfaces (low, medium or high)
  • Specifically formulated for total bacterial count and for detecting yeast and moulds, enterobacteria, E. coli and coliform
  • Easily used and transported away from the laboratory to control the surfaces of production plants
  • Easy to use thanks to the screw cap