Cell viability assay kits, OZBlue
Leverancier: OZ BIOSCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BL00025 BL00100
OZBIBL00025EA 164.01 EUR
OZBIBL00025 OZBIBL00100
Cell viability assay kits, OZBlue
Testen Cellulaire assays
OZBlue cell viability kit is a ready to use assay system based on the fluorimetric/colorimetric quantification of metabolic activity in living cells. Resazurin dye is used as an indicator of cell viability.
  • Non toxic: Allows additional analysis on proliferating cells (mRNA, cytogenetic, apoptosis, immunophenotyping) and continuous monitoring of cultures over the time
  • High sensitivity: More sensitive than (³H) thymidine incorporation and tetrazolium reduction assay (MTT, XTT)
  • Simple protocol: No need for washing or extraction procedures
  • Versatile: Can be used on cell lines, primary cells, bacteria, fungi
  • Flexible: Colorimetric and fluorescence detection allows compatibility with flow cytometry, plate readers and microscopy detection
Store at +4 °C.