Pipette stands, hanging
Leverancier: IBI SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PST-50 PST-51
SHETPST-50EA 86.4 EUR
SHETPST-50 SHETPST-51
Pipette stands, hanging
Pipethouders
These four-place pipette holders have an additional place for a mechanical pipette filler. The pipette rack is constructed of durable cast acrylic for years of reliable use, and the fluorescent orange color will add colour to the lab. These are orientented such that they can be placed on the right or left side of the fume hood.
  • Holds four standard pipettes and a media filler pipette
  • Attaches to hanging surface with powerful double-sided tape
The PST-50 is oriented so that the pipette filler will be located on the right side, while the pipette filler of PST-51 will be located on the left side.