Connector kits for EASY-Column™ capillary HPLC columns
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SC602 SC601 SC600 SC603
HYPESC602EA 0 EUR
HYPESC602 HYPESC601 HYPESC600 HYPESC603
Connector kits for EASY-Column™ capillary HPLC columns
Fittingen en connectoren
Use these connector kits, sleeves, and tees with EASY-Column™ capillary columns.
  • Zero dead volume connector kits
  • Available for one- and two-column configurations
  • Simple, reproducible setup for on-line LC/MS
Each column delivers excellent chromatographic performance on any nanoscale LC system. At flow rates of nl/min versus ml/min, the EASY-Column offers higher sensitivity with greater signal-to-noise ratio compared to traditional electrospray.