Thermal transfer printer labels, type B-486, MetaLabel
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
020785 020782
BRDY020785EA 366.31 EUR
BRDY020785 BRDY020782
Thermal transfer printer labels, type B-486, MetaLabel
Etiketten
Printable, ultra aggressive metalised polyester labels for thermal transfer printers.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.