Thermal transfer printer labels, type B-480, MetaLabel
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
104185 104186 104195 104197
BRDY104185EA 562.02 EUR
BRDY104185 BRDY104186 BRDY104195 BRDY104197
Thermal transfer printer labels, type B-480, MetaLabel
Etiketten
These labels coupled with Brady's IP™ Thermal Transfer Printing System, are the only metallic product available for hassle-free printing.
Supplied with an industrial-strength adhesive, as well as the Mondo Bondo™ adhesive, you can trust these labels in dirty, oily environments, and you can easily wipe clean the label front.