Avantor® Alltima HP, HPLC columns, HILIC, 5 µm
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HICH86465EA 1100 EUR
HICH86465 HICH86464 HICH86466 86479/N 86480/N
Avantor® Alltima HP, HPLC columns, HILIC, 5 µm
Kolommen voor chromatografie
Avantor® Alltima HP columns features hydrophilic interaction chromatography that uses small amounts of water for increased sensitivity with microbore applications.
  • High resolution for complex samples
  • High purity silica
  • Excellent column stability
These columns combine the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
These are widely used for polar analytes that are difficult to retain by reversed-phase.
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage Lage impact op de fabricage
Oppervlakte 230 m²/g
USP code L3
Milieu certificering ISO 14001:2015 (530840 UM15) ISO 14001:2015 (530840 UM15)