Reference materials, acid number standards, total acid number (TAN)
Leverancier: LGC Standards PROMOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CON150-800-051 CON150-800-151 CON150-800-455
PROOCON150-800-051EA 0 EUR
PROOCON150-800-051 PROOCON150-800-151 PROOCON150-800-455
Reference materials, acid number standards, total acid number (TAN)
Standaarden Standaarden voor de petroleumindustrie
Acid number reference materials are intended for use in the determination of acid number (AN) in petroleum products in accordance with ASTM D664/IP 177 (by potentiometric titration) or ASTM D974 (by colour-indicator titration).
Certificaten: GMP/GLP, ISO 9001, ISO/IEC 17025 and ISO/IEC 17043 certified.