Total organic carbon standards, according to ISO/CEN EN1484, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
76067-250ML-F 76067-500ML-F
76067-250ML-FEA 116 EUR
76067-250ML-F 76067-500ML-F
Total organic carbon standards, according to ISO/CEN EN1484, Supelco®
Standaarden Standaarden voor milieuanalyse
  • Stabilized
  • Limited shelf life, expiry date on the label
Opgelet: Storage temperature: 2…8 °C