Ring lights, EasyLED
Leverancier: SCHOTT AG Lighting and imaging
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
600200 600300
631-0884EA 600 EUR
631-0884 630-1200
Ring lights, EasyLED
Lampen Ringlichten
The EasyLED product line integrates advanced LED illumination with control functions in a single compact device. It is the ideal standard illumination system for routine inspections and education.
  • Powerful illuminance
  • Compact and ergonomic
  • Integrated controller
  • Mountable direct to the microscope objective
  • International clip plug system
The EasyLED ring light offers homogeneous and shadow-free illumination in a robust metal housing and well designed heat sink that enables maximum brightness and long lifetime of 50000 hours. The multi LED light head offers an attractive alternative to conventional cold light sources with fiber optics.
The EasyLED ring light plus has the additional advantage of controllable segments, which enables new contrasting methods. The integrated 'jog dial' element allows an easy and intuitive switching between the different illumination modes as well as rotation of the segments in both directions.