Labels for LaserTab® laser printer, yype B-773
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
014393 029751 014394 014395 014396 014936 029688 029749 029750 029839 029860 029864
BRDY014393EA 455.12 EUR
BRDY014393 BRDY029751 BRDY014394 BRDY014395 BRDY014396 BRDY014936 BRDY029688 BRDY029749 BRDY029750 BRDY029839 BRDY029860 BRDY029864
Labels for LaserTab® laser printer, yype B-773
Etiketten
Blank, printable polyester labels for Laser printers.
  • Designed to be printed with a Laser or InkJet printer
  • Available blank or pre-printed with a template or header
  • Supplied on 203,2×254 mm or A4 209,8×297,2 mm sheets
This material is a UL recognised component and CSA accepted material when printed with designated laser printer toners. See UL file MH17154 and CSA Acceptance Record LS41833 for specific details.