Indoor Outdoor Vinyl Labels, M211 and M210
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
142807 142794 142801 142810 142809 142808 142804 142806 142805 142802 142800 142799 142798 142797 142796 142795 142803 142811 139733 139732 139734 139735 139736 139737 139738 139739 139741 139742 139744 139745
BRDY142807EA 35.14 EUR
BRDY142807 BRDY142794 BRDY142801 811-0164 BRDY142809 BRDY142808 BRDY142804 BRDY142806 BRDY142805 BRDY142802 BRDY142800 116-0633 BRDY142798 116-0632 BRDY142796 811-0163 BRDY142803 BRDY142811 BRDY139733 BRDY139732 BRDY139734 BRDY139735 BRDY139736 BRDY139737 BRDY139738 BRDY139739 BRDY139741 BRDY139742 BRDY139744 BRDY139745
Indoor Outdoor Vinyl Labels, M211 and M210
Etiketten
These B-595 vinyl labels will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics.
  • Average outdoor durability: 8 to 10 years
  • Gloss finish
  • For smooth and textured/rough surfaces
The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.