Self-laminating Polyester Cryogenic Laboratory Labels, M211 and M210
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
110933 110932 110931
817-0114EA 50.04 EUR
817-0114 817-0113 817-0112
Self-laminating Polyester Cryogenic Laboratory Labels, M211 and M210
Etiketten
Printable high-performance laboratory self-laminating polyester.
  • Matt finish
  • Ultra-thin construction prevents jamming in racks and centrifuges
Suitable for liquid nitrogen application if material is wrapped around itself.