Mop heads, polyester, quiltec I
Leverancier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MHQT0150 MHQT0100
568-0910EA 290 EUR
568-0910 568-0909
Mop heads, polyester, quiltec I
Moppen
Mop heads are constructed from a two-ply quilted 100% polyester on the outer face with a synthetic fibrefill core.
  • Designed for single use
  • Multi-layer sewn lamination
  • Mop heads are compatible with a wide range of chemicals
Highly sorbent mop heads are suitable for the application of disinfectants and removal of light soil, residues and biofilms.